Learn More
Medchemexpress LLC Indacaterol maleate | 753498-25-8 | C28H32N2O7 | 500 MG
SDP
Catalog No. 5000354953
Encompass_Preferred
Indacaterol maleate is an orally active, ultra-long-acting β2 adrenergic receptor (ADRB2) agonist. It inhibits NF-κB activity in a β-arrestin2-dependent manner, which helps prevent further lung damage and improves lung function in chronic obstructive pulmonary disorder (COPD). It can also be utilized in cardiovascular disease research.
  • Orally active, ultra-long-acting β2 adrenergic receptor (ADRB2) agonist.
  • Inhibits NF-κB activity in a β-arrestin2-dependent manner.
  • Helps prevent further lung damage.
  • Improves lung function in chronic obstructive pulmonary disorder (COPD).
  • Can be used in cardiovascular disease research.
Catalog No. 50-003-54953 Supplier Medchemexpress LLC Supplier No. HY14299A500MG
Indacaterol maleate is an orally active, ultra-long-acting β2 adrenergic receptor (ADRB2) agonist. It inhibits NF-κB activity in a β-arrestin2-dependent manner, which helps prevent further lung damage and improves lung function in chronic obstructive pulmonary disorder (COPD). It can also be utilized in cardiovascular disease research.
  • Orally active, ultra-long-acting β2 adrenergic receptor (ADRB2) agonist.
  • Inhibits NF-κB activity in a β-arrestin2-dependent manner.
  • Helps prevent further lung damage.
  • Improves lung function in chronic obstructive pulmonary disorder (COPD).
  • Can be used in cardiovascular disease research.