Learn More
Medchemexpress LLC Indacaterol maleate | 753498-25-8 | MFCD20526769 | 99.6% | C28H32N2O7 | 10MG
SDP
Catalog No. 5000261021
Encompass_Preferred
Indacaterol maleate is the maleate salt of indacaterol, an ultra-long-acting β2-adrenergic receptor agonist. This product is supplied as an analytical reference standard for research and laboratory use and is intended for assay development, method validation, and quality control (not for human use).
  • High-purity reference material (HPLC 99.62%).
  • Suitable for assay development and method validation.
  • Intended for research and analytical use only.
  • Supplied as a stable solid for standard preparation.
  • Identified by CAS 753498-25-8 for unambiguous reference.
Catalog No. 50-002-61021 Supplier Medchemexpress LLC Supplier No. HY14299AR10MG
Indacaterol maleate is the maleate salt of indacaterol, an ultra-long-acting β2-adrenergic receptor agonist. This product is supplied as an analytical reference standard for research and laboratory use and is intended for assay development, method validation, and quality control (not for human use).
  • High-purity reference material (HPLC 99.62%).
  • Suitable for assay development and method validation.
  • Intended for research and analytical use only.
  • Supplied as a stable solid for standard preparation.
  • Identified by CAS 753498-25-8 for unambiguous reference.