Learn More
Medchemexpress LLC Indacaterol maleate | 753498-25-8 | MFCD20526769 | 99.8% | 508.56 g/mol | C28H32N2O7 | 5 MG
SDP
Catalog No. 5000221623
Encompass_Preferred
Indacaterol maleate is the maleate salt of indacaterol, an ultra-long-acting β2-adrenergic receptor agonist. Supplied as an analytical standard, this solid reference material is intended for research use in assay development, method validation, and quality control testing.
  • High purity (99.8%).
  • Suitable for assay development and method validation.
  • Confirmed identity by LC-MS consistent with structure.
  • Stable solid form for accurate weighing and storage.
  • Includes CAS 753498-25-8 and molecular weight 508.56 g/mol.
Catalog No. 50-002-21623 Supplier Medchemexpress LLC Supplier No. HY14299AR5MG
Indacaterol maleate is the maleate salt of indacaterol, an ultra-long-acting β2-adrenergic receptor agonist. Supplied as an analytical standard, this solid reference material is intended for research use in assay development, method validation, and quality control testing.
  • High purity (99.8%).
  • Suitable for assay development and method validation.
  • Confirmed identity by LC-MS consistent with structure.
  • Stable solid form for accurate weighing and storage.
  • Includes CAS 753498-25-8 and molecular weight 508.56 g/mol.