Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indacaterol Maleate 98.0+%, TCI America™
SDP

Catalog No. I0927100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0927100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0927100MG Supplier TCI America Supplier No. I0927100MG

missing translation for 'validMainframeMsg'

Specifications

CAS 753498-25-8
Melting Point 186°C
Color White-Yellow
Molecular Formula C28H32N2O7
MDL Number MFCD20526769,MFCD18782702
Quantity 100 mg
Synonym (R)-5-[2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxyquinolin-2(1H)-one Maleate
InChI Key IREJFXIHXRZFER-UHFFFAOYNA-N
SMILES OC(=O)C=CC(O)=O.CCC1=C(CC)C=C2CC(CC2=C1)NCC(O)C1=C2C=CC(=O)NC2=C(O)C=C1
IUPAC Name 5-{2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl}-8-hydroxy-1,2-dihydroquinolin-2-one; but-2-enedioic acid
Molecular Weight (g/mol) 508.57
PubChem CID 66713197
Formula Weight 508.57
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Indacaterol Maleate
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.