Learn More
Cayman Chemical Indapamide-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2934053-07-1, 1 mg
SDP
Catalog No. 502949059
Encompass_Preferred
An internal standard for the quantification of indapamide by GC- or LC-MS
Catalog No. 50-294-9059 Supplier Cayman Chemical Supplier No. 342441 MG
An internal standard for the quantification of indapamide by GC- or LC-MS