Learn More
Cayman Chemical IndapamIde-13C-d3 500ug
SDP
Catalog No. 502948201
Encompass_Preferred
An internal standard for the quantification of indapamide by GC- or LC-MS
Catalog No. 50-294-8201 Supplier Cayman Chemical Supplier No. 34244500 UG
An internal standard for the quantification of indapamide by GC- or LC-MS