Learn More
Apexbio Technology LLC Indapamide 26807-65-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093102
Encompass_Preferred
Indapamide (CAS 26807-65-8) is a sulphonamide-derived diuretic structurally distinct from thiazides It functions by inhibiting sodium reabsorption primarily at the distal convoluted tubules of the nephron resulting in increased excretion of sodium and water This action reduces plasma volume and peripheral vascular resistance contributing to its antihypertensive effects Indapamide is also studied for its utility in models of decompensated cardiac failure due to its natriuretic and vasodilatory properties making it relevant for research into cardiovascular disease and fluid balance regulation
Catalog No. 50-000-93102 Supplier Apexbio Technology LLC Supplier No. B19535.1
Indapamide (CAS 26807-65-8) is a sulphonamide-derived diuretic structurally distinct from thiazides It functions by inhibiting sodium reabsorption primarily at the distal convoluted tubules of the nephron resulting in increased excretion of sodium and water This action reduces plasma volume and peripheral vascular resistance contributing to its antihypertensive effects Indapamide is also studied for its utility in models of decompensated cardiac failure due to its natriuretic and vasodilatory properties making it relevant for research into cardiovascular disease and fluid balance regulation