Learn More
Medchemexpress LLC HY-B0259 500mg Medchemexpress, Indapamide CAS:26807-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0259500MG
Encompass_Preferred
Medchemexpress, HY-B0259 500mg Indapamide CAS:26807-65-8 Indapamide is an orally active sulphonamide diuretic agent, that can reduce blood pressure by decreasing vascular reactivity and peripheral vascular resistance. Indapamide is also can reduce left ventricular hypertrophy[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2429