Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indapamide

Catalog No. AAJ6384603
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAJ6384603

Supplier: Thermo Scientific Chemicals J6384603

Only null left
Add to Cart

CAS: 26807-65-8 | C16H16ClN3O3S | 365.83 g/mol

Used as an antihypertensive. Diuretic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used as an antihypertensive
  • Diuretic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26807-65-8
Molecular Formula C16H16ClN3O3S
Molecular Weight (g/mol) 365.83
MDL Number MFCD00079375
InChI Key NDDAHWYSQHTHNT-UHFFFAOYNA-N
Synonym indapamide, noranat, veroxil, tertensif, arifon, fludex, lozol, indaflex, bajaten, indamol
PubChem CID 3702
ChEBI CHEBI:5893
IUPAC Name 4-chloro-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)-3-sulfamoylbenzamide
SMILES CC1CC2=CC=CC=C2N1NC(=O)C1=CC=C(Cl)C(=C1)S(N)(=O)=O

Specifications

Melting Point 160°C to 162°C
Color White
Quantity 1 g
Merck Index 14,4935
Solubility Information Soluble in ethanol; Insoluble in water
Formula Weight 365.83
Physical Form Powder
Chemical Name or Material Indapamide
EINECSNumber 248-012-7
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.