Learn More
Medchemexpress LLC Indazole | 271-44-3 | 99.8% | 118.14 | 1 ML
SDP
Catalog No. 5000404068
Encompass_Preferred
Indazole, also called isoindazole, is a heterocyclic aromatic organic compound. Its derivatives exhibit a broad spectrum of biological activities. It is utilized in research concerning cancer, neurological disorders, cardiovascular diseases, and gastrointestinal diseases.
  • Derivatives display a broad variety of biological activities.
  • Used in research for cancer, neurological disorders, cardiovascular diseases, and gastrointestinal diseases.
  • Compounds show potential anticancer activity.
  • Demonstrates potent activities against neurological disorders.
  • Shows promising results in treating various gastrointestinal disorders.
Catalog No. 50-004-04068 Supplier Medchemexpress LLC Supplier No. HY4029410MM/1ML
Indazole, also called isoindazole, is a heterocyclic aromatic organic compound. Its derivatives exhibit a broad spectrum of biological activities. It is utilized in research concerning cancer, neurological disorders, cardiovascular diseases, and gastrointestinal diseases.
  • Derivatives display a broad variety of biological activities.
  • Used in research for cancer, neurological disorders, cardiovascular diseases, and gastrointestinal diseases.
  • Compounds show potential anticancer activity.
  • Demonstrates potent activities against neurological disorders.
  • Shows promising results in treating various gastrointestinal disorders.