Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indazole-3-carboxylic acid | 4498-67-3 | 162.15 | 25 G
SDP

Catalog No. 5000448118
Encompass_Preferred

Indazole-3-carboxylic acid is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is intended for research use only and is not for sale to patients.

  • Purity: 99.39%.
  • Molecular weight: 162.15.
  • Formula: C8H6N2O2.
  • Appearance: Solid.
  • Color: Light yellow to orange.
  • Solubility: Soluble in DMSO (≥ 200 mg/mL or 1233.43 mM). Users are advised to use newly opened DMSO due to its hygroscopic nature, which significantly impacts solubility.
  • Storage: Store at room temperature for 3 years. In solvent, store at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-004-48118 Supplier Medchemexpress LLC Supplier No. 25GHYW002518
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indazole-3-carboxylic acid is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is intended for research use only and is not for sale to patients.

  • Purity: 99.39%.
  • Molecular weight: 162.15.
  • Formula: C8H6N2O2.
  • Appearance: Solid.
  • Color: Light yellow to orange.
  • Solubility: Soluble in DMSO (≥ 200 mg/mL or 1233.43 mM). Users are advised to use newly opened DMSO due to its hygroscopic nature, which significantly impacts solubility.
  • Storage: Store at room temperature for 3 years. In solvent, store at -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.