Learn More
TARGETMOL CHEMICALS INC INDAZOLE-CL 5MG
SDP
Catalog No. 502580748
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Indazole-Cl is a selective ERß agonist and a selective estrogen receptor modifier (SERM). purity: 98%
Catalog No. 50-258-0748 Supplier TARGETMOL CHEMICALS INC Supplier No. T255345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Indazole-Cl is a selective ERß agonist and a selective estrogen receptor modifier (SERM). purity: 98%