Learn More
Medchemexpress LLC Indazole-Cl | 848142-62-1 | 98.19% | 260.68 | 1 MG
SDP
Catalog No. 5000352058
Encompass_Preferred
Indazole-Cl (Ind-Cl) is an Estrogen receptor (ER)-β-specific agonist with inflammatory effect. It inhibits cyclooxygenase-2 expression reduction induced by hypoxia and also inhibits ROS production. Indazole-Cl inhibits cell migration and invasion increased by hypoxia. It is a potent inhibitor of hypoxia-induced inflammation in vascular smooth muscle cells (VSMCs).
  • Estrogen receptor (ER)-β-specific agonist
  • Inflammatory effect
  • Inhibits cyclooxygenase-2 expression reduction induced by hypoxia
  • Inhibits ROS production
  • Inhibits cell migration and invasion by hypoxia
  • Potent inhibitor of hypoxia-induced inflammation in vascular smooth muscle cells (VSMCs)
Catalog No. 50-003-52058 Supplier Medchemexpress LLC Supplier No. HY1370041MG
Indazole-Cl (Ind-Cl) is an Estrogen receptor (ER)-β-specific agonist with inflammatory effect. It inhibits cyclooxygenase-2 expression reduction induced by hypoxia and also inhibits ROS production. Indazole-Cl inhibits cell migration and invasion increased by hypoxia. It is a potent inhibitor of hypoxia-induced inflammation in vascular smooth muscle cells (VSMCs).
  • Estrogen receptor (ER)-β-specific agonist
  • Inflammatory effect
  • Inhibits cyclooxygenase-2 expression reduction induced by hypoxia
  • Inhibits ROS production
  • Inhibits cell migration and invasion by hypoxia
  • Potent inhibitor of hypoxia-induced inflammation in vascular smooth muscle cells (VSMCs)