Learn More
Medchemexpress LLC Indeno[1,2,3-cd]pyrene | 193-39-5 | 99.7% | 276.33 | 1 MG
SDP
Catalog No. 5000401976
Encompass_Preferred
Indeno[1,2,3-cd]pyrene is a polycyclic aromatic hydrocarbon that exhibits moderate cytotoxicity in human pulmonary alveolar epithelial cells (HPAEpiC). It enhances allergic lung inflammation via the aryl hydrocarbon receptor.
  • Exhibits moderate cytotoxicity in human pulmonary alveolar epithelial cells (HPAEpiC)
  • Enhances allergic lung inflammation via the aryl hydrocarbon receptor
Catalog No. 50-004-01976 Supplier Medchemexpress LLC Supplier No. HY234991MG
Add to Cart
Add to Cart
Indeno[1,2,3-cd]pyrene is a polycyclic aromatic hydrocarbon that exhibits moderate cytotoxicity in human pulmonary alveolar epithelial cells (HPAEpiC). It enhances allergic lung inflammation via the aryl hydrocarbon receptor.
  • Exhibits moderate cytotoxicity in human pulmonary alveolar epithelial cells (HPAEpiC)
  • Enhances allergic lung inflammation via the aryl hydrocarbon receptor