Learn More
Medchemexpress LLC Indeno[1,2,3-cd]pyrene-d12 | 203578-33-0 | 288.40 | 5 MG
SDP
Catalog No. 5000402140
Encompass_Preferred
Indeno[1,2,3-cd]pyrene-d12 is a deuterium-labeled compound intended for research use only. This compound is valuable as a tracer and an internal standard in quantitative analysis, particularly with NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules can influence their pharmacokinetic and metabolic profiles, making this compound significant for various research applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-02140 Supplier Medchemexpress LLC Supplier No. HY23499S5MG
Indeno[1,2,3-cd]pyrene-d12 is a deuterium-labeled compound intended for research use only. This compound is valuable as a tracer and an internal standard in quantitative analysis, particularly with NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, such as deuterium, into drug molecules can influence their pharmacokinetic and metabolic profiles, making this compound significant for various research applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can affect pharmacokinetic and metabolic profiles