Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Indenyl)titanium(IV) Trichloride 98.0+%, TCI America™
SDP

Catalog No. I06461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06461G Supplier TCI America Supplier No. I06461G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 84365-55-9
Molecular Formula C9H7Cl3Ti
Molecular Weight (g/mol) 269.372
InChI Key MDTDQDVMQBTXST-UHFFFAOYSA-K
Synonym trichloro indenyl titanium iv, trichloro 1h-inden-1-yl titanium, acmc-20ajew, indenyltrichlorotitanium, trichloroindenyltitanium, indenyl trichlorotitanium, indenyltitanium trichloride
PubChem CID 15302996
IUPAC Name 1H-inden-1-ide;trichlorotitanium(1+)
SMILES [CH-]1C=CC2=CC=CC=C21.Cl[Ti+](Cl)Cl

Specifications

Melting Point 162°C
Color Black-Blue
Quantity 1 g
UN Number 3261
Formula Weight 269.37
Percent Purity ≥98.0% (W,T)
Physical Form Crystalline Powder
Chemical Name or Material (Indenyl)titanium(IV) Trichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.