Learn More
Strem, An Ascensus Company CAS# 84365-55-9. 5g. Indenyltitanium trichloride, 99%. MFCD00269849
SDP
Catalog No. 5090110760
Encompass
CAS# 84365-55-9. 5g. Indenyltitanium trichloride, 99%. MFCD00269849. Molecular Weight: 269.39. Molecular Formula: C9H7TiCl3. Color/form: purple xtl. Strem# 22-0950. http://www.strem.com/catalog/v/22-0950/
Catalog No. 50-901-10760 Supplier Strem, An Ascensus Company Supplier No. 2209505G
CAS# 84365-55-9. 5g. Indenyltitanium trichloride, 99%. MFCD00269849. Molecular Weight: 269.39. Molecular Formula: C9H7TiCl3. Color/form: purple xtl. Strem# 22-0950. http://www.strem.com/catalog/v/22-0950/