Learn More
Vernier™ Independence of Motion Accessory
SDPSCI_ED
Catalog No. S15992ND Shop All Vernier Software Products
Change view
Click to view available options
Includes:
Built-in level
Catalog No. Includes
S15992ND Built-in level
1 options
Catalog No. S15992ND Supplier Vernier™ Supplier No. IOMVPL
Add to Cart
Add to Cart
Includes
Independence of Motion Accessory
The Independence of Motion Accessory enables the Vernier Projectile Launcher to perform the classic experiment where one ball is dropped as another is projected horizontally. The balls strike the floor simultaneously.
A built-in level makes leveling the accessory fast and easy. One ball is placed in the barrel, which causes a pin to extend. A second, drilled ball is placed on the extended pin. When the first ball is ejected from the Projectile Launcher, the pin retracts and the second ball falls. Both metal and plastic balls are provided.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
For Use With (Equipment) Go Direct™ Projectile Launcher, Vernier™ Projectile Launcher
Includes Built-in level