Learn More
Hampton Research HR2-944-01/Index #01 (A1), 185 ml - 0.1 M Citric acid pH 3.5, 2.0 M Ammonium sulfate
SDP
Catalog No. 502174
Encompass
HR2-944-01/Index #01 (A1), 185 ml - 0.1 M Citric acid pH 3.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-217-4 Supplier Hampton Research Supplier No. HR294401
HR2-944-01/Index #01 (A1), 185 ml - 0.1 M Citric acid pH 3.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.