Learn More
Hampton Research HR2-944-02/Index #02 (A2), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 2.0 M Ammonium sulfate
SDP
Catalog No. 502175
Encompass
HR2-944-02/Index #02 (A2), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-217-5 Supplier Hampton Research Supplier No. HR294402
HR2-944-02/Index #02 (A2), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.