Learn More
Hampton Research HR2-944-03/Index #03 (A3), 185 ml - 0.1 M BIS-TRIS pH 5.5, 2.0 M Ammonium sulfate
SDP
Catalog No. 502176
Encompass
HR2-944-03/Index #03 (A3), 185 ml - 0.1 M BIS-TRIS pH 5.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-217-6 Supplier Hampton Research Supplier No. HR294403
HR2-944-03/Index #03 (A3), 185 ml - 0.1 M BIS-TRIS pH 5.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.