Learn More
Hampton Research HR2-944-06/Index #06 (A6), 185 ml - 0.1 M Tris pH 8.5, 2.0 M Ammonium sulfate
SDP
Catalog No. 502179
Encompass
HR2-944-06/Index #06 (A6), 185 ml - 0.1 M Tris pH 8.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.
Catalog No. 50-217-9 Supplier Hampton Research Supplier No. HR294406
HR2-944-06/Index #06 (A6), 185 ml - 0.1 M Tris pH 8.5, 2.0 M Ammonium sulfate. Please note items from this supplier are non-returnable.