Learn More
Hampton Research HR2-944-11/Index #11 (A11), 185 ml - 0.1 M HEPES pH 7.5, 3.0 M Sodium chloride
SDP
Catalog No. 502184
Encompass
HR2-944-11/Index #11 (A11), 185 ml - 0.1 M HEPES pH 7.5, 3.0 M Sodium chloride. Please note items from this supplier are non-returnable.
Catalog No. 50-218-4 Supplier Hampton Research Supplier No. HR294411
HR2-944-11/Index #11 (A11), 185 ml - 0.1 M HEPES pH 7.5, 3.0 M Sodium chloride. Please note items from this supplier are non-returnable.