Learn More
Hampton Research HR2-944-13/Index #13 (B1), 185 ml - 0.1 M BIS-TRIS pH 5.5, 0.3 M Magnesium formate dihydrate
SDP
Catalog No. 502186
Encompass
HR2-944-13/Index #13 (B1), 185 ml - 0.1 M BIS-TRIS pH 5.5, 0.3 M Magnesium formate dihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-218-6 Supplier Hampton Research Supplier No. HR294413
HR2-944-13/Index #13 (B1), 185 ml - 0.1 M BIS-TRIS pH 5.5, 0.3 M Magnesium formate dihydrate. Please note items from this supplier are non-returnable.