Learn More
Hampton Research HR2-944-18/Index #18 (B6), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.9
SDP
Catalog No. 502191
Encompass
HR2-944-18/Index #18 (B6), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.
Catalog No. 50-219-1 Supplier Hampton Research Supplier No. HR294418
HR2-944-18/Index #18 (B6), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate/Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.