Learn More
Hampton Research HR2-944-31/Index #31 (C7), 185 ml - 0.8 M Potassium sodium tartrate tetrahydrate, 0.1 M Tris pH 8.5, 0.5% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502204
Encompass
HR2-944-31/Index #31 (C7), 185 ml - 0.8 M Potassium sodium tartrate tetrahydrate, 0.1 M Tris pH 8.5, 0.5% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-220-4 Supplier Hampton Research Supplier No. HR294431
HR2-944-31/Index #31 (C7), 185 ml - 0.8 M Potassium sodium tartrate tetrahydrate, 0.1 M Tris pH 8.5, 0.5% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.