Learn More
Hampton Research HR2-944-32/Index #32 (C8), 185 ml - 1.0 M Ammonium sulfate, 0.1 M BIS-TRIS pH 5.5, 1% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502205
Encompass
HR2-944-32/Index #32 (C8), 185 ml - 1.0 M Ammonium sulfate, 0.1 M BIS-TRIS pH 5.5, 1% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-220-5 Supplier Hampton Research Supplier No. HR294432
HR2-944-32/Index #32 (C8), 185 ml - 1.0 M Ammonium sulfate, 0.1 M BIS-TRIS pH 5.5, 1% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.