Learn More
Hampton Research HR2-944-33/Index #33 (C9), 185 ml - 1.1 M Sodium malonate pH 7.0, 0.1 M HEPES pH 7.0, 0.5% v/v Jeffamine ED-2001 pH 7.0
SDP
Catalog No. 502206
Encompass
HR2-944-33/Index #33 (C9), 185 ml - 1.1 M Sodium malonate pH 7.0, 0.1 M HEPES pH 7.0, 0.5% v/v Jeffamine ED-2001 pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-220-6 Supplier Hampton Research Supplier No. HR294433
HR2-944-33/Index #33 (C9), 185 ml - 1.1 M Sodium malonate pH 7.0, 0.1 M HEPES pH 7.0, 0.5% v/v Jeffamine ED-2001 pH 7.0. Please note items from this supplier are non-returnable.