Learn More
Hampton Research HR2-944-34/Index #34 (C10), 185 ml - 1.0 M Succinic acid pH 7.0, 0.1 M HEPES pH 7.0, 1% w/v Polyethylene glycol monomethyl ether 2,000
SDP
Catalog No. 502207
Encompass
HR2-944-34/Index #34 (C10), 185 ml - 1.0 M Succinic acid pH 7.0, 0.1 M HEPES pH 7.0, 1% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-220-7 Supplier Hampton Research Supplier No. HR294434
HR2-944-34/Index #34 (C10), 185 ml - 1.0 M Succinic acid pH 7.0, 0.1 M HEPES pH 7.0, 1% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.