Learn More
Hampton Research HR2-944-35/Index #35 (C11), 185 ml - 1.0 M Ammonium sulfate, 0.1 M HEPES pH 7.0, 0.5% w/v Polyethylene glycol 8,000
SDP
Catalog No. 502208
Encompass
HR2-944-35/Index #35 (C11), 185 ml - 1.0 M Ammonium sulfate, 0.1 M HEPES pH 7.0, 0.5% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.
Catalog No. 50-220-8 Supplier Hampton Research Supplier No. HR294435
HR2-944-35/Index #35 (C11), 185 ml - 1.0 M Ammonium sulfate, 0.1 M HEPES pH 7.0, 0.5% w/v Polyethylene glycol 8,000. Please note items from this supplier are non-returnable.