Learn More
Hampton Research HR2-944-36/Index #36 (C12), 185 ml - 15% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.0, 2% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502209
Encompass
HR2-944-36/Index #36 (C12), 185 ml - 15% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.0, 2% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-220-9 Supplier Hampton Research Supplier No. HR294436
HR2-944-36/Index #36 (C12), 185 ml - 15% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.0, 2% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.