Learn More
Hampton Research HR2-944-39/Index #39 (D3), 185 ml - 0.1 M HEPES pH 7.0, 30% v/v Jeffamine ED-2001 pH 7.0
SDP
Catalog No. 502212
Encompass
HR2-944-39/Index #39 (D3), 185 ml - 0.1 M HEPES pH 7.0, 30% v/v Jeffamine ED-2001 pH 7.0. Please note items from this supplier are non-returnable.
Catalog No. 50-221-2 Supplier Hampton Research Supplier No. HR294439
HR2-944-39/Index #39 (D3), 185 ml - 0.1 M HEPES pH 7.0, 30% v/v Jeffamine ED-2001 pH 7.0. Please note items from this supplier are non-returnable.