Learn More
Hampton Research HR2-944-40/Index #40 (D4), 185 ml - 0.1 M Citric acid pH 3.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502213
Encompass
HR2-944-40/Index #40 (D4), 185 ml - 0.1 M Citric acid pH 3.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-221-3 Supplier Hampton Research Supplier No. HR294440
HR2-944-40/Index #40 (D4), 185 ml - 0.1 M Citric acid pH 3.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.