Learn More
Hampton Research HR2-944-41/Index #41 (D5), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502214
Encompass
HR2-944-41/Index #41 (D5), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-221-4 Supplier Hampton Research Supplier No. HR294441
HR2-944-41/Index #41 (D5), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.