Learn More
Hampton Research HR2-944-43/Index #43 (D7), 185 ml - 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502216
Encompass
HR2-944-43/Index #43 (D7), 185 ml - 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-221-6 Supplier Hampton Research Supplier No. HR294443
HR2-944-43/Index #43 (D7), 185 ml - 0.1 M BIS-TRIS pH 6.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.