Learn More
Hampton Research HR2-944-44/Index #44 (D8), 185 ml - 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502217
Encompass
HR2-944-44/Index #44 (D8), 185 ml - 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-221-7 Supplier Hampton Research Supplier No. HR294444
HR2-944-44/Index #44 (D8), 185 ml - 0.1 M HEPES pH 7.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.