Learn More
Hampton Research HR2-944-45/Index #45 (D9), 185 ml - 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502218
Encompass
HR2-944-45/Index #45 (D9), 185 ml - 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-221-8 Supplier Hampton Research Supplier No. HR294445
HR2-944-45/Index #45 (D9), 185 ml - 0.1 M Tris pH 8.5, 25% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.