Learn More
Hampton Research HR2-944-46/Index #46 (D10), 185 ml - 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502219
Encompass
HR2-944-46/Index #46 (D10), 185 ml - 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-221-9 Supplier Hampton Research Supplier No. HR294446
HR2-944-46/Index #46 (D10), 185 ml - 0.1 M BIS-TRIS pH 6.5, 20% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.