Learn More
Hampton Research HR2-944-47/Index #47 (D11), 185 ml - 0.1 M BIS-TRIS pH 6.5, 28% w/v Polyethylene glycol monomethyl ether 2,000
SDP
Catalog No. 502220
Encompass
HR2-944-47/Index #47 (D11), 185 ml - 0.1 M BIS-TRIS pH 6.5, 28% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-222-0 Supplier Hampton Research Supplier No. HR294447
HR2-944-47/Index #47 (D11), 185 ml - 0.1 M BIS-TRIS pH 6.5, 28% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.