Learn More
Hampton Research HR2-944-49/Index #49 (E01), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M BIS-TRIS pH 6.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 502222
Encompass
HR2-944-49/Index #49 (E01), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M BIS-TRIS pH 6.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-222-2 Supplier Hampton Research Supplier No. HR294449
HR2-944-49/Index #49 (E01), 185 ml - 0.2 M Calcium chloride dihydrate, 0.1 M BIS-TRIS pH 6.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.