Learn More
Hampton Research HR2-944-50/Index #50 (E02), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 5.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 502223
Encompass
HR2-944-50/Index #50 (E02), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 5.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-222-3 Supplier Hampton Research Supplier No. HR294450
HR2-944-50/Index #50 (E02), 185 ml - 0.2 M Ammonium acetate, 0.1 M BIS-TRIS pH 5.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.