Learn More
Hampton Research HR2-944-53/Index #53 (E05), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol
SDP
Catalog No. 502226
Encompass
HR2-944-53/Index #53 (E05), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-222-6 Supplier Hampton Research Supplier No. HR294453
HR2-944-53/Index #53 (E05), 185 ml - 0.2 M Ammonium acetate, 0.1 M Tris pH 8.5, 45% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.