Learn More
Hampton Research HR2-944-54/Index #54 (E06), 185 ml - 0.05 M Calcium chloride dihydrate, 0.1 M BIS-TRIS pH 6.5, 30% v/v Polyethylene glycol monomethyl ether 550
SDP
Catalog No. 502227
Encompass
HR2-944-54/Index #54 (E06), 185 ml - 0.05 M Calcium chloride dihydrate, 0.1 M BIS-TRIS pH 6.5, 30% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.
Catalog No. 50-222-7 Supplier Hampton Research Supplier No. HR294454
HR2-944-54/Index #54 (E06), 185 ml - 0.05 M Calcium chloride dihydrate, 0.1 M BIS-TRIS pH 6.5, 30% v/v Polyethylene glycol monomethyl ether 550. Please note items from this supplier are non-returnable.