Learn More
Hampton Research HR2-944-59/Index #59 (E11), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.1 M HEPES pH 7.5, 22% w/v Polyacrylic acid sodium salt 5,100
SDP
Catalog No. 502232
Encompass
HR2-944-59/Index #59 (E11), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.1 M HEPES pH 7.5, 22% w/v Polyacrylic acid sodium salt 5,100. Please note items from this supplier are non-returnable.
Catalog No. 50-223-2 Supplier Hampton Research Supplier No. HR294459
HR2-944-59/Index #59 (E11), 185 ml - 0.02 M Magnesium chloride hexahydrate, 0.1 M HEPES pH 7.5, 22% w/v Polyacrylic acid sodium salt 5,100. Please note items from this supplier are non-returnable.