Learn More
Hampton Research HR2-944-60/Index #60 (E12), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 20% w/v Polyvinylpyrrolidone K 15
SDP
Catalog No. 502233
Encompass
HR2-944-60/Index #60 (E12), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 20% w/v Polyvinylpyrrolidone K 15. Please note items from this supplier are non-returnable.
Catalog No. 50-223-3 Supplier Hampton Research Supplier No. HR294460
HR2-944-60/Index #60 (E12), 185 ml - 0.01 M Cobalt(II) chloride hexahydrate, 0.1 M Tris pH 8.5, 20% w/v Polyvinylpyrrolidone K 15. Please note items from this supplier are non-returnable.