Learn More
Hampton Research HR2-944-61/Index #61 (F01), 185 ml - 0.2 M L-Proline, 0.1 M HEPES pH 7.5, 10% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502234
Encompass
HR2-944-61/Index #61 (F01), 185 ml - 0.2 M L-Proline, 0.1 M HEPES pH 7.5, 10% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-223-4 Supplier Hampton Research Supplier No. HR294461
HR2-944-61/Index #61 (F01), 185 ml - 0.2 M L-Proline, 0.1 M HEPES pH 7.5, 10% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.