Learn More
Hampton Research HR2-944-62/Index #62 (F02), 185 ml - 0.2 M Trimethylamine N-oxide dihydrate, 0.1 M Tris pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 2,000
SDP
Catalog No. 502235
Encompass
HR2-944-62/Index #62 (F02), 185 ml - 0.2 M Trimethylamine N-oxide dihydrate, 0.1 M Tris pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.
Catalog No. 50-223-5 Supplier Hampton Research Supplier No. HR294462
HR2-944-62/Index #62 (F02), 185 ml - 0.2 M Trimethylamine N-oxide dihydrate, 0.1 M Tris pH 8.5, 20% w/v Polyethylene glycol monomethyl ether 2,000. Please note items from this supplier are non-returnable.