Learn More
Hampton Research HR2-944-63/Index #63 (F03), 185 ml - 5% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.0, 10% w/v Polyethylene glycol monomethyl ether 5,000
SDP
Catalog No. 502236
Encompass
HR2-944-63/Index #63 (F03), 185 ml - 5% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.0, 10% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.
Catalog No. 50-223-6 Supplier Hampton Research Supplier No. HR294463
HR2-944-63/Index #63 (F03), 185 ml - 5% v/v Tacsimate pH 7.0, 0.1 M HEPES pH 7.0, 10% w/v Polyethylene glycol monomethyl ether 5,000. Please note items from this supplier are non-returnable.