Learn More
Hampton Research HR2-944-64/Index #64 (F04), 185 ml - 0.005 M Cobalt(II) chloride, 0.005 M Nickel(II) chloride, 0.005 M Cadmium chloride, 0.005 M Magnesium chloride, 0.1 M HEPES pH 7.5, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502237
Encompass
HR2-944-64/Index #64 (F04), 185 ml - 0.005 M Cobalt(II) chloride, 0.005 M Nickel(II) chloride, 0.005 M Cadmium chloride, 0.005 M Magnesium chloride, 0.1 M HEPES pH 7.5, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-223-7 Supplier Hampton Research Supplier No. HR294464
HR2-944-64/Index #64 (F04), 185 ml - 0.005 M Cobalt(II) chloride, 0.005 M Nickel(II) chloride, 0.005 M Cadmium chloride, 0.005 M Magnesium chloride, 0.1 M HEPES pH 7.5, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.