Learn More
Hampton Research HR2-944-65/Index #65 (F05), 185 ml - 0.1 M Ammonium acetate, 0.1 M BIS-TRIS pH 5.5, 17% w/v Polyethylene glycol 10,000
SDP
Catalog No. 502238
Encompass
HR2-944-65/Index #65 (F05), 185 ml - 0.1 M Ammonium acetate, 0.1 M BIS-TRIS pH 5.5, 17% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.
Catalog No. 50-223-8 Supplier Hampton Research Supplier No. HR294465
HR2-944-65/Index #65 (F05), 185 ml - 0.1 M Ammonium acetate, 0.1 M BIS-TRIS pH 5.5, 17% w/v Polyethylene glycol 10,000. Please note items from this supplier are non-returnable.